missing translation for 'onlineSavingsMsg'
Learn More

4-Isocyanato-1-methyl-1H-pyrazole, 95%, ACROS Organics™

Brand:  Acros Organics 431140010

Code : 29

Additional Details : CAS Number : 1174064-57-3 Weight : 0.05100kg Transport : UN number : 2206 Chem class : 6.1 Pack group : III


Quantity 1g
Packaging Glass bottle

 View more versions of this product

Product Code. 10442980

  • £125.50 / 1g


Estimated Shipment
20-06-2018
Add to basket


Specifications

Chemical Identifiers

4-Isocyanato-1-methyl-1H-pyrazole
94.0
95%
C5H5N3O
MFCD14706124
1174064-57-3
100.0
95%
123.12

Specifications

4-Isocyanato-1-methyl-1H-pyrazole
1174064-57-3
100.0
95%
C5H5N3O
MFCD14706124
Glass bottle
Yellow
1g
94.0
Liquid, May Contain Fine Suspension
95%
123.12
Authentic
203.0° to 205.0°C
71°C
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 4-Isocyanato-1-methyl-1H-pyrazole

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3

Hazard Statement
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H331-Toxic if inhaled.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P311-Call a POISON CENTER/doctor/ .
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .

SDS