Learn More
Alfa Aesar™ 4-Isochromanone, 95%
Brand: Alfa Aesar™ H61539.03
Code : NEW
Additional Details : CAS Number : 20924-56-5 Weight : 0.00100kg
Quantity | 1g |
---|
Chemical Identifiers
4-Isochromanone | |
20924-56-5 | |
148.16 | |
MFCD09744058 |
Specifications
4-Isochromanone | |
20924-56-5 | |
95% | |
MFCD09744058 |
1g | |
Solid | |
148.16 |