Learn More
4-IPP, Tocris Bioscience™
Inhibitor of macrophage migration inhibitory factor (MIF); suicide substrate
Brand:  Tocris (Bio-Techne) 342950
Code : Z1
Additional Details : Weight : 0.01000kg
Quantity 50mg
 View more versions of this product
Product Code. 13462994
  • £529.12 / 50mg
Estimated Shipment
07-03-2019
Add to basket
Description
Description
Inhibitor of macrophage migration inhibitory factor (MIF). Acts as a suicide substrate to inactivate MIF catalytic and biological functions; inhibits MIF-associated liver enzyme activity in vivo. Also inhibits migration and anchorage independence of human lung adenocarcinoma cell lines in vitro.
Specifications
Chemical Identifiers
41270-96-6
282.084
4-ipp, d0w1wf, 4-ipp hplc
4-iodo-6-phenylpyrimidine
C10H7IN2
ZTCJXHNJVLUUMR-UHFFFAOYSA-N
817368
C1=CC=C(C=C1)C2=CC(=NC=N2)I
Specifications
4-IPP
41270-96-6
4-ipp, d0w1wf, 4-ipp hplc
C1=CC=C(C=C1)C2=CC(=NC=N2)I
282.084
282.08
50mg
C10H7IN2
ZTCJXHNJVLUUMR-UHFFFAOYSA-N
4-iodo-6-phenylpyrimidine
817368
>99%
Videos