missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Iodotetrahydro-2H-pyran, ≥97%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Specifications
CAS Min % | 94.5 |
CAS Max % | 100.0 |
Assay Percent Range | 95% |
Packaging | Amber Glass Bottle |
Quantity | 1 g |
Formula Weight | 212.03 |
Percent Purity | 95% |
Chemical Name or Material | 4-Iodotetrahydro-2H-pyran |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'