missing translation for 'onlineSavingsMsg'
Learn More

4-Iodophenyl isothiocyanate, 97%, Maybridge™

£7.27 - £23.90

Chemical Identifiers

CAS 2059-76-9
Molecular Formula C7H4INS
Molecular Weight (g/mol) 261.08
MDL Number MFCD00041091
InChI Key SNLGBRZZFRSXHA-UHFFFAOYSA-N
Synonym 4-iodophenyl isothiocyanate, 4-iodophenylisothiocyanate, 1-iodo-4-isothiocyanato-benzene, benzene, 1-iodo-4-isothiocyanato, 4-ipitc, acmc-20amg6, 4-iodobenzenisothiocyanate, 1-isothiocyanato-4-iodobenzene, #, benzene,1-iodo-4-isothiocyanato
PubChem CID 74938
IUPAC Name 1-iodo-4-isothiocyanatobenzene
SMILES C1=CC(=CC=C1N=C=S)I
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10009591 SDS View Documents Maybridge
S00350DA
1g Amber Glass Bottle £7.27
1g View alternative products
Save Up to 
»
Estimated Shipment
17-08-2020
Please to purchase this item. Need a web account? Register with us today!
10009601 SDS View Documents Maybridge
S00350EA
10g Amber Glass Bottle £23.90
10g View alternative products
Save Up to 
»
Estimated Shipment
17-08-2020
Please to purchase this item. Need a web account? Register with us today!
Specifications

Chemical Identifiers

2059-76-9
261.08
SNLGBRZZFRSXHA-UHFFFAOYSA-N
74938
C1=CC(=CC=C1N=C=S)I
C7H4INS
MFCD00041091
4-iodophenyl isothiocyanate, 4-iodophenylisothiocyanate, 1-iodo-4-isothiocyanato-benzene, benzene, 1-iodo-4-isothiocyanato, 4-ipitc, acmc-20amg6, 4-iodobenzenisothiocyanate, 1-isothiocyanato-4-iodobenzene, #, benzene,1-iodo-4-isothiocyanato
1-iodo-4-isothiocyanatobenzene

Specifications

4-Iodophenyl isothiocyanate
1g
97.0
C7H4INS
MFCD00041091
SNLGBRZZFRSXHA-UHFFFAOYSA-N
1-iodo-4-isothiocyanatobenzene
74938
97%
Amber Glass Bottle
2059-76-9
97%
IC6H4NCS
4-iodophenyl isothiocyanate, 4-iodophenylisothiocyanate, 1-iodo-4-isothiocyanato-benzene, benzene, 1-iodo-4-isothiocyanato, 4-ipitc, acmc-20amg6, 4-iodobenzenisothiocyanate, 1-isothiocyanato-4-iodobenzene, #, benzene,1-iodo-4-isothiocyanato
C1=CC(=CC=C1N=C=S)I
261.08
261.08
Safety and Handling

Safety and Handling

Product Identifier
  • 4-Iodophenyl isothiocyanate

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.