missing translation for 'onlineSavingsMsg'
Learn More

4-(Iodomethyl)tetrahydro-2h-pyran, 97%, Maybridge

Brand:  Maybridge CC29940DE

Code : 31

Additional Details : CAS Number : 101691-94-5 Weight : 0.05500kg Transport : UN number : 3265 Chem class : 8 Pack group : II

Quantity 5g
Packaging Amber Glass Bottle

 View more versions of this product

Product Code. 10570554

  • £185.38 / 5g


Estimated Shipment date
from Supplier 01-06-2018
Add to basket

Specifications

Chemical Identifiers

4-(Iodomethyl)tetrahydro-2H-pyran
97%
226.05
97%
C6H11IO

Specifications

4-(Iodomethyl)tetrahydro-2H-pyran
Amber Glass Bottle
97%
226.05
101691-94-5
97%
C6H11IO
5g
Videos
SDS