missing translation for 'onlineSavingsMsg'
Learn More

4-Iodobenzoyl chloride, 98%, ACROS Organics™

Brand:  Acros Organics 412390050

Code : 29

Additional Details : CAS Number : 1711-02-0 Weight : 0.06000kg Transport : UN number : 3261 Chem class : 8 Pack group : III


Packaging Glass bottle
Quantity 5g

 View more versions of this product

Product Code. 10053923

  • £25.00 / 5g

Estimated Shipment: 11-10-2021

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Chemical Identifiers

1711-02-0
266.46
NJAKCIUOTIPYED-UHFFFAOYSA-N
74373
C1=CC(=CC=C1C(=O)Cl)I
C7H4ClIO
MFCD00001058
p-iodobenzoyl chloride, benzoyl chloride, 4-iodo, benzoyl chloride, p-iodo, 4-iodobenzoic acid chloride, 4-iodobenzoylchloride, 4-iodo-benzoyl chloride, 4iodobenzoyl chloride, 4-iodo-benzoylchlorid, rho-iodobenzoyl chloride, 4-iodo benzoyl chloride
4-iodobenzoyl chloride

Specifications

4-Iodobenzoyl chloride
Glass bottle
62°C to 66°C
98%
97.5
97.5% min. (GC)
IC6H4COCl
09, 367
NJAKCIUOTIPYED-UHFFFAOYSA-N
4-iodobenzoyl chloride
74373
Crystalline Powder and Chunks or Crystalline Mass
Authentic
Yellow to Beige
5g
1711-02-0
100.0
C7H4ClIO
MFCD00001058
p-iodobenzoyl chloride, benzoyl chloride, 4-iodo, benzoyl chloride, p-iodo, 4-iodobenzoic acid chloride, 4-iodobenzoylchloride, 4-iodo-benzoyl chloride, 4iodobenzoyl chloride, 4-iodo-benzoylchlorid, rho-iodobenzoyl chloride, 4-iodo benzoyl chloride
C1=CC(=CC=C1C(=O)Cl)I
266.46
266.46
98%
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 4-Iodobenzoyl chloride

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B

Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS Documents
Certificates