missing translation for 'onlineSavingsMsg'
Learn More

4-Iodobenzotrifluoride, 98%, Thermo Scientific Chemicals

Catalog Number p-7081974
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
455-13-0
C7H4F3I
272.009
MFCD00039398
SKGRFPGOGCHDPC-UHFFFAOYSA-N
4-iodobenzotrifluoride, 1-iodo-4-trifluoromethyl benzene, p-iodobenzotrifluoride, benzene, 1-iodo-4-trifluoromethyl, 4-trifluoromethyl iodobenzene, 4-iodo-alpha,alpha,alpha-trifluorotoluene, alpha,alpha,alpha-trifluoro-4-iodotoluene, 4-iodobenztrifluoride, 4-iodobenzo trifluoride, 4-iodo-1-trifluoromethylbenzene
67993
1-iodo-4-(trifluoromethyl)benzene
C1=CC(=CC=C1C(F)(F)F)I
This item is not returnable. View return policy
455-13-0
C7H4F3I
272.009
MFCD00039398
SKGRFPGOGCHDPC-UHFFFAOYSA-N
4-iodobenzotrifluoride, 1-iodo-4-trifluoromethyl benzene, p-iodobenzotrifluoride, benzene, 1-iodo-4-trifluoromethyl, 4-trifluoromethyl iodobenzene, 4-iodo-alpha,alpha,alpha-trifluorotoluene, alpha,alpha,alpha-trifluoro-4-iodotoluene, 4-iodobenztrifluoride, 4-iodobenzo trifluoride, 4-iodo-1-trifluoromethylbenzene
67993
1-iodo-4-(trifluoromethyl)benzene
C1=CC(=CC=C1C(F)(F)F)I

4-Iodobenzotrifluoride may be employed as substrate with electron-deficient aromatic ring, during Mizoroki-Heck reaction with acrylic acid, to afford 4-trifluoromethylcinnnamic acid. It undergoes aminocarbonylation in DMF using phosphoryl chloride to give N,N-dimethyl-(4-trifluoromethyl)benzamide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Iodobenzotrifluoride may be employed as substrate with electron-deficient aromatic ring, during Mizoroki-Heck reaction with acrylic acid, to afford 4-trifluoromethylcinnnamic acid. It undergoes aminocarbonylation in DMF using phosphoryl chloride to give N,N-dimethyl-(4-trifluoromethyl)benzamide.

Solubility
Insoluble in water.

Notes
Light sensitive. Incompatible with strong oxidizing agents. Store in dark.
TRUSTED_SUSTAINABILITY
Melting Point -8°C
Density 1.851
Boiling Point 68°C to 70°C (15 mmHg)
Flash Point 68°C (154°F)
Refractive Index 1.518
Quantity 1 g
Beilstein 1944013
Sensitivity Light sensitive
Solubility Information Insoluble in water.
Formula Weight 272.01
Percent Purity 98%
Chemical Name or Material 4-Iodobenzotrifluoride
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'