missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Iodo-5-methyl-isoxazole, 97%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
5 g
Specifications
CAS Min % | 96.5 |
CAS Max % | 100.0 |
Assay Percent Range | 97% |
Quantity | 5 g |
IUPAC Name | 6-[(5Z)-5-({5-cyano-2-[3-(ethoxycarbonyl)piperidin-1-yl]-4-methyl-6-oxo-1-propyl-1,6-dihydropyridin-3-yl}methylidene)-4-oxo-2-sulfanylidene-1,3-thiazolidin-3-yl]hexanoic acid |
Formula Weight | 208.98 |
Percent Purity | 97% |
Chemical Name or Material | 4-Iodo-5-methyl-isoxazole |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'