missing translation for 'onlineSavingsMsg'
Learn More

4-Iodo-3-methyl-5-phenylisoxazole, 97%, Thermo Scientific™

Catalog Number p-2701866
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
16114-53-7
C10H8INO
285.084
MFCD03086126
RLZGZJOMCXKYIW-UHFFFAOYSA-N
4-iodo-3-methyl-5-phenylisoxazole, isoxazole,4-iodo-3-methyl-5-phenyl, 3-methyl-5-phenyl-4-iodoisoxazole, 4-iodo-3-methylisoxazol-5-yl benzene, 4-iodo-3-methyl-1,2-oxazol-5-yl benzene
2776520
4-iodo-3-methyl-5-phenyl-1,2-oxazole
CC1=NOC(=C1I)C2=CC=CC=C2
This item is not returnable. View return policy
16114-53-7
C10H8INO
285.084
MFCD03086126
RLZGZJOMCXKYIW-UHFFFAOYSA-N
4-iodo-3-methyl-5-phenylisoxazole, isoxazole,4-iodo-3-methyl-5-phenyl, 3-methyl-5-phenyl-4-iodoisoxazole, 4-iodo-3-methylisoxazol-5-yl benzene, 4-iodo-3-methyl-1,2-oxazol-5-yl benzene
2776520
4-iodo-3-methyl-5-phenyl-1,2-oxazole
CC1=NOC(=C1I)C2=CC=CC=C2
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 285.08
Percent Purity 97%
Chemical Name or Material 4-Iodo-3-methyl-5-phenylisoxazole
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'