missing translation for 'onlineSavingsMsg'
Learn More

4-Iodo-2-methylaniline, 98+%, Thermo Scientific™

Catalog Number p-7058266
Click to view available options
Quantity:
25 g
This item is not returnable. View return policy
13194-68-8
C7H8IN
233.052
MFCD00025299
BGKLFAQCHHCZRZ-UHFFFAOYSA-N
2-amino-5-iodotoluene, 4-iodo-2-methylbenzenamine, 4-iodo-o-toluidine, benzenamine, 4-iodo-2-methyl, 4-iodo-2-methyl-phenylamine, 2-methyl-4-iodoaniline, 4-iodo-2-methylphenylamine, 4-iodo-2-methyl-aniline, aniline, 4-iodo-2-methyl, aurora 1126
83221
4-iodo-2-methylaniline
CC1=C(C=CC(=C1)I)N
This item is not returnable. View return policy
13194-68-8
C7H8IN
233.052
MFCD00025299
BGKLFAQCHHCZRZ-UHFFFAOYSA-N
2-amino-5-iodotoluene, 4-iodo-2-methylbenzenamine, 4-iodo-o-toluidine, benzenamine, 4-iodo-2-methyl, 4-iodo-2-methyl-phenylamine, 2-methyl-4-iodoaniline, 4-iodo-2-methylphenylamine, 4-iodo-2-methyl-aniline, aniline, 4-iodo-2-methyl, aurora 1126
83221
4-iodo-2-methylaniline
CC1=C(C=CC(=C1)I)N

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 84°C to 88°C
Quantity 25 g
UN Number UN2811
Beilstein 2353618
Sensitivity Light sensitive
Formula Weight 233.05
Percent Purity ≥98%
Chemical Name or Material 4-Iodo-2-methylaniline
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'