Learn More
4-IBP, Tocris Bioscience™
σ ligand, σ1 > σ2
Brand:  Tocris (Bio-Techne) 074810
Code : Z1
Additional Details : Weight : 0.01000kg
Quantity 10mg
 View more versions of this product
Product Code. 13487903
  • £114.75 / 10mg
Estimated Shipment
03-10-2018
Add to basket
Description
Description
Combines high affinity for σ1 and moderate affinity for σ2 sites. Centrally active following systemic administration in vivo.
Specifications
Chemical Identifiers
155798-08-6
420.294
N-(N-Benzylpiperidin-4-yl)-4-iodobenzamide
C19H21IN2O
HELCSESNNDZLFM-UHFFFAOYSA-N
132995
Specifications
4-IBP
>99%
N-(N-Benzylpiperidin-4-yl)-4-iodobenzamide
HELCSESNNDZLFM-UHFFFAOYSA-N
N-(1-benzylpiperidin-4-yl)-4-iodobenzamide
132995
155798-08-6
C19H21IN2O
10mg
C1CN(CCC1NC(=O)C2=CC=C(C=C2)I)CC3=CC=CC=C3
420.294
420.29
Videos