missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxythiophenol, 97%, Thermo Scientific Chemicals

Catalog Number p-7044079
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
637-89-8
C6H6OS
126.17
MFCD00004850
BXAVKNRWVKUTLY-UHFFFAOYSA-N
4-mercaptophenol, 4-hydroxythiophenol, 4-hydroxybenzenethiol, p-hydroxythiophenol, phenol, 4-mercapto, p-mercaptophenol, thiohydroquinone, monothiohydroquinone, 4-hydroxy thiophenol, hydroquinone, monothio
240147
4-sulfanylphenol
OC1=CC=C(S)C=C1
This item is not returnable. View return policy
637-89-8
C6H6OS
126.17
MFCD00004850
BXAVKNRWVKUTLY-UHFFFAOYSA-N
4-mercaptophenol, 4-hydroxythiophenol, 4-hydroxybenzenethiol, p-hydroxythiophenol, phenol, 4-mercapto, p-mercaptophenol, thiohydroquinone, monothiohydroquinone, 4-hydroxy thiophenol, hydroquinone, monothio
240147
4-sulfanylphenol
OC1=CC=C(S)C=C1

4-Hydroxythiophenol is used to improve charge separation in PbSe quantum dots (QDs) as sensitizers for TiO2. It helps in the activation of neocarzinostatin-chromophore. Also used as a resin monomer in the study of the different gold nanocrystal core-resin shell structures

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Hydroxythiophenol is used to improve charge separation in PbSe quantum dots (QDs) as sensitizers for TiO2. It helps in the activation of neocarzinostatin-chromophore. Also used as a resin monomer in the study of the different gold nanocrystal core-resin shell structures

Solubility
Soluble in water.

Notes
Air Sensitive. Store at 4°C. Incompatible with air and oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 33°C to 37°C
Density 1.255
Boiling Point 256°C
Flash Point 110°C (230°F)
Odor Unpleasant
Quantity 5 g
UN Number UN1759
Beilstein 2039306
Sensitivity Air Sensitive
Solubility Information Soluble in water.
Formula Weight 126.18
Percent Purity 97%
Chemical Name or Material 4-Hydroxythiophenol
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'