missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxyquinazoline 98%, ACROS Organics™

Brand:  Acros Organics 121880050

Code : 29

Additional Details : CAS Number : 491-36-1 Weight : 0.05500kg


Quantity 5g
Packaging Glass bottle

 View more versions of this product

Product Code. 10487192

  • £15.00 / 5g


Estimated Shipment
13-02-2018
Add to basket


Specifications

Chemical Identifiers

4-Hydroxyquinazoline
491-36-1
100.0
98%
146.15
4-Quinazolinol
98%
97.5
98%
C8H6N2O
MFCD00511302

Specifications

4-Hydroxyquinazoline
215.0° to 219.0°C
98%
97.5
Authentic
Crystalline Powder
98%
146.15
24, 143
Off-white to light beige
5g
491-36-1
100.0
Glass bottle
98%
C8H6N2O
MFCD00511302
4-Quinazolinol
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 4-Hydroxyquinazoline

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS