missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxyphenylsulfur pentafluoride, 95%, Thermo Scientific™

Catalog Number 10822071
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy
CAS 774-94-7
Assay Percent Range 95%
Packaging Amber Glass Bottle
Molecular Formula C6H5F5OS
MDL Number MFCD03788516
Quantity 1 g
Synonym 4-pentafluorothio phenol, 4-pentafluorosulfanyl phenol, 4-hydroxyphenylsulfur pentafluoride, 4-pentafluoro-$l^ 6-sulfanyl phenol, 4-pentafluoro-lambda∼6∼-sulfanyl phenol, sulfur,pentafluoro 4-hydroxyphenyl-, oc-6-21-9ci, 4-pentafluorosulfanylphenol, 4-pentafluorosulfur phenol, 4-pentafluorosulphanyl phenol, 4-pentafluorothio vi phenol
InChI Key XHJLGVIUMCBMHL-UHFFFAOYSA-N
SMILES OC1=CC=C(C=C1)S(F)(F)(F)(F)F
IUPAC Name 4-(pentafluoro-$l^{6}-sulfanyl)phenol
Molecular Weight (g/mol) 220.16
PubChem CID 2779203
Formula Weight 220.16
Percent Purity 95%
Chemical Name or Material 4-Hydroxyphenylsulfur pentafluoride
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'