missing translation for 'onlineSavingsMsg'
Learn More

4-(Hydroxymethyl)-1,3-dioxolan-2-one, 90%, Thermo Scientific Chemicals

Catalog Number 10491050
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle

 

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS 931-40-8
CAS Min % 88.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.4000g/mL
Elemental Analysis 36.6 to 44.7% C, 4.6 to 5.6% H, 48.8 to 59.6% O
Boiling Point 142.5°C (1.0 mmHg)
Flash Point 110°C
Infrared Spectrum Authentic
Assay Percent Range 90%
Packaging Glass bottle
Molecular Formula C4H6O4
Refractive Index 1.4597
MDL Number MFCD00085561
Quantity 25 g
Specific Gravity 1.4
Synonym 4-hydroxymethyl-1,3-dioxolan-2-one, glycerol carbonate, hydroxymethyl dioxolanone, glycerol 1,2-carbonate, 1,3-dioxolan-2-one, 4-hydroxymethyl, 1,3-dioxolan-2-one,4-hydroxymethyl, glycerin carbonate, glycerol1,2-carbonate, 4-hydroxymethyl-1,3-dioxolan-2-on
InChI Key JFMGYULNQJPJCY-UHFFFAOYNA-N
SMILES OCC1COC(=O)O1
IUPAC Name 4-(hydroxymethyl)-1,3-dioxolan-2-one
Molecular Weight (g/mol) 118.09
PubChem CID 97944
Formula Weight 118.09
Percent Purity 90%
Physical Form Liquid
Chemical Name or Material 4-(Hydroxymethyl)-1, 3-dioxolan-2-one
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'