missing translation for 'onlineSavingsMsg'
Learn More

4'-Hydroxydiclofenac, Thermo Scientific Chemicals

Catalog Number 15426199
Click to view available options
Quantity:
5 mg

 

A Cytochrome P450 2C9 (CYP2C9) metabolite of diclofenac

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS 64118-84-9
Color Yellow
Molecular Formula C14H11Cl2NO3
MDL Number MFCD01671980
Quantity 5 mg
UN Number UN2811
Beilstein 4198042
Synonym 4'-hydroxydiclofenac, 4'-hydroxy diclofenac, o-2,6-dichloro-4-hydroxyanilino phenyl acetic acid, unii-njf5o599ef, 4'-oh dcf, 2-2,6-dichloro-4-hydroxyphenyl amino phenyl acetic acid, 4′-ohd, njf5o599ef, diclofenac, 4′-hydroxy, benzeneacetic acid, 2-2,6-dichloro-4-hydroxyphenyl amino
Solubility Information Soluble in DMSO, ethanol or methanol
InChI Key KGVXVPRLBMWZLG-UHFFFAOYSA-N
SMILES C1=CC=C(C(=C1)CC(=O)O)NC2=C(C=C(C=C2Cl)O)Cl
IUPAC Name 2-[2-(2,6-dichloro-4-hydroxyanilino)phenyl]acetic acid
Molecular Weight (g/mol) 312.146
PubChem CID 116545
ChEBI CHEBI:59613
Formula Weight 312.15
Physical Form Powder
Chemical Name or Material 4'-Hydroxydiclofenac
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'