missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxycoumarin, 98%, Thermo Scientific Chemicals

Catalog Number 10568480
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
Plastic bottle
This item is not returnable. View return policy
1076-38-6
C9H6O3
162.14
MFCD00006856
OWBBAPRUYLEWRR-UHFFFAOYSA-N
4-hydroxycoumarin, 4-coumarinol, 4-hydroxy-2h-chromen-2-one, benzotetronic acid, coumarin, 4-hydroxy, 2h-1-benzopyran-2-one, 4-hydroxy, 4-hydroxy-2h-1-benzopyran-2-one, 4-hydroxy-1-benzopyran-2-one, 4-hydroxy-chromen-2-one, unii-x954zll2rd
54682930
CHEBI:40070
OC1=CC(=O)C2=CC=CC=C2O1
This item is not returnable. View return policy
1076-38-6
C9H6O3
162.14
MFCD00006856
OWBBAPRUYLEWRR-UHFFFAOYSA-N
4-hydroxycoumarin, 4-coumarinol, 4-hydroxy-2h-chromen-2-one, benzotetronic acid, coumarin, 4-hydroxy, 2h-1-benzopyran-2-one, 4-hydroxy, 4-hydroxy-2h-1-benzopyran-2-one, 4-hydroxy-1-benzopyran-2-one, 4-hydroxy-chromen-2-one, unii-x954zll2rd
54682930
CHEBI:40070
OC1=CC(=O)C2=CC=CC=C2O1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 210°C to 215°C
Color Colorless to Yellow
Loss on Drying 0.5% max.
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 5 g
Beilstein 17, 488
Solubility Information Solubility in water: practically insoluble in water. Other solubilities: soluble in hot water,soluble in alcohol,acetone and ether
IUPAC Name 4-hydroxychromen-2-one
Formula Weight 162.14
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 4-Hydroxycoumarin, 98%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'