missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxycoumarin, 98%, Acros Organics

£13.00 - £95.20

Chemical Identifiers

CAS 1076-38-6
Molecular Formula C9H6O3
Molecular Weight (g/mol) 162.14
MDL Number MFCD00006856
InChI Key VXIXUWQIVKSKSA-UHFFFAOYSA-N
Synonym 4-hydroxycoumarin, 4-coumarinol, 4-hydroxy-2h-chromen-2-one, benzotetronic acid, coumarin, 4-hydroxy, 2h-1-benzopyran-2-one, 4-hydroxy, 4-hydroxy-2h-1-benzopyran-2-one, 4-hydroxy-1-benzopyran-2-one, 4-hydroxy-chromen-2-one, unii-x954zll2rd
PubChem CID 54682930
ChEBI CHEBI:40070
IUPAC Name 4-hydroxychromen-2-one
SMILES C1=CC=C2C(=C1)C(=CC(=O)O2)O
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10568480 SDS View Documents Acros Organics
121100050
5g Glass bottle £13.00
5g View alternative products
Save Up to 
»
EU Stock 9 Please to purchase this item. Need a web account? Register with us today!
10578480 SDS View Documents Acros Organics
121101000
100g Plastic bottle £21.90
100g View alternative products
Save Up to 
»
EU Stock 7 Please to purchase this item. Need a web account? Register with us today!
10182770 SDS View Documents Acros Organics
121105000
500g Plastic bottle £95.20
500g View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 10-03-2020
Please to purchase this item. Need a web account? Register with us today!
Specifications

Chemical Identifiers

1076-38-6
162.14
VXIXUWQIVKSKSA-UHFFFAOYSA-N
54682930
4-hydroxychromen-2-one
C9H6O3
MFCD00006856
4-hydroxycoumarin, 4-coumarinol, 4-hydroxy-2h-chromen-2-one, benzotetronic acid, coumarin, 4-hydroxy, 2h-1-benzopyran-2-one, 4-hydroxy, 4-hydroxy-2h-1-benzopyran-2-one, 4-hydroxy-1-benzopyran-2-one, 4-hydroxy-chromen-2-one, unii-x954zll2rd
CHEBI:40070
C1=CC=C2C(=C1)C(=CC(=O)O2)O

Specifications

4-Hydroxycoumarin
0.5% max.
0.25% max.
210°C to 215°C
98%
97.5
98%
MFCD00006856
4-hydroxycoumarin, 4-coumarinol, 4-hydroxy-2h-chromen-2-one, benzotetronic acid, coumarin, 4-hydroxy, 2h-1-benzopyran-2-one, 4-hydroxy, 4-hydroxy-2h-1-benzopyran-2-one, 4-hydroxy-1-benzopyran-2-one, 4-hydroxy-chromen-2-one, unii-x954zll2rd
VXIXUWQIVKSKSA-UHFFFAOYSA-N
4-hydroxychromen-2-one
54682930
162.14
98%
Authentic
Glass bottle
Colorless to Yellow
5g
1076-38-6
100.0
C9H6O3
17, 488
Solubility in water: practically insoluble in water. Other solubilities: soluble in hot water,soluble in alcohol,acetone and ether
C1=CC=C2C(=C1)C(=CC(=O)O2)O
162.14
CHEBI:40070
Liquid