missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Hydroxybutyric acid hydrazide, 98%

Brand:  Alfa Aesar™ A12351.14

Code : NEW

Additional Details : CAS Number : 3879-08-1 Weight : 0.02500kg

Quantity 25g

 View more versions of this product

Product Code. 11409483

  • £87.80 / 25g


Estimated Shipment date
from Supplier 31-01-2018
Add to basket

Specifications

Chemical Identifiers

4-Hydroxybutyric acid hydrazide
HO(CH2)3CONHNH2
MFCD00051702
4-Hydroxybutanoic hydrazide
3879-08-1
118.14
1749234

Specifications

4-Hydroxybutyric acid hydrazide
25g
98%
118.14
1749234
91° to 95°C
3879-08-1
HO(CH2)3CONHNH2
MFCD00051702
4-Hydroxybutanoic hydrazide
Videos
SDS