missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4'-Hydroxybiphenyl-4-carboxamidoxime, 97%

Brand:  Alfa Aesar™ H58802.06

Code : NEW

Additional Details : Weight : 0.00500kg

Quantity 5g

 View more versions of this product

Product Code. 15455008

  • £825.74 / 5g


Estimated Shipment date
from Supplier 12-06-2018
Add to basket

Specifications

Chemical Identifiers

4'-Hydroxybiphenyl-4-carboxamidoxime
228.25
MFCD22581245

Specifications

4'-Hydroxybiphenyl-4-carboxamidoxime
5g
97%
MFCD22581245
Pale brown
Solid
228.25
Videos
SDS