missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxybenzyl alcohol, 97%, ACROS Organics™

£25.14 - £271.70

Chemical Identifiers

CAS 623-05-2
Molecular Formula C7H8O2
Molecular Weight (g/mol) 124.14
InChI Key BVJSUAQZOZWCKN-UHFFFAOYSA-N
Synonym 4-hydroxybenzyl alcohol, 4-hydroxymethyl phenol, p-hydroxybenzyl alcohol, p-methylolphenol, 4-methylolphenol, 4-hydroxybenzenemethanol, benzenemethanol, 4-hydroxy, p-hydroxymethyl phenol, gastrodigenin, 4-hydroxybenzylalcohol
PubChem CID 125
ChEBI CHEBI:67410
IUPAC Name 4-(hydroxymethyl)phenol
SMILES C1=CC(=CC=C1CO)O
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10784431 SDS View Documents Acros Organics
121030250
25g Glass bottle £25.14
25g View alternative products
Save Up to 
»
EU Stock 9 Please to purchase this item. Need a web account? Register with us today!
10336900 SDS View Documents Acros Organics
121031000
100g Glass bottle £78.13
100g View alternative products
Save Up to 
»
EU Stock 12 Please to purchase this item. Need a web account? Register with us today!
11934671 SDS View Documents Acros Organics
121035000
500g Glass bottle £271.70
500g View alternative products
Save Up to 
»
EU Stock 3 Please to purchase this item. Need a web account? Register with us today!
Specifications

Chemical Identifiers

623-05-2
124.14
4-hydroxybenzyl alcohol, 4-hydroxymethyl phenol, p-hydroxybenzyl alcohol, p-methylolphenol, 4-methylolphenol, 4-hydroxybenzenemethanol, benzenemethanol, 4-hydroxy, p-hydroxymethyl phenol, gastrodigenin, 4-hydroxybenzylalcohol
CHEBI:67410
C1=CC(=CC=C1CO)O
C7H8O2
BVJSUAQZOZWCKN-UHFFFAOYSA-N
125
4-(hydroxymethyl)phenol

Specifications

4-Hydroxybenzyl alcohol
Glass bottle
110.0°C to 112.0°C
623-05-2
C7H8O2
06,897
Solubility in water: soluble
C1=CC(=CC=C1CO)O
124.14
CHEBI:67410
Crystalline Powder
Authentic
Beige to Pink
25g
96% min. (HPLC)
HOC6H4CH2OH
4-hydroxybenzyl alcohol, 4-hydroxymethyl phenol, p-hydroxybenzyl alcohol, p-methylolphenol, 4-methylolphenol, 4-hydroxybenzenemethanol, benzenemethanol, 4-hydroxy, p-hydroxymethyl phenol, gastrodigenin, 4-hydroxybenzylalcohol
BVJSUAQZOZWCKN-UHFFFAOYSA-N
4-(hydroxymethyl)phenol
125
124.14
97%
Safety and Handling

Safety and Handling

Product Identifier
  • 4-Hydroxybenzyl alcohol

Signal Word
  • Warning

Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.