missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxybenzenesulfonamide, 97%, ACROS Organics™

£16.50 - £186.80

Chemical Identifiers

CAS 1576-43-8
Molecular Formula C6H7NO3S
Molecular Weight (g/mol) 1730.19
MDL Number MFCD00059190
InChI Key DIRCLGLKRZLKHG-UHFFFAOYSA-N
Synonym p-hydroxybenzenesulfonamide, p-phenolsulfonamide, benzenesulfonamide, 4-hydroxy, 4-sulfamoylphenol, benzenesulfonamide, p-hydroxy, 4-hydroxyphenylsulfonamide, unii-1iit7r7vpj, p-hydroxybenzenesulphonamide, 4-hydroxybenzenesulphonamide, 4-hydroxybenzene-1-sulfonamide
PubChem CID 74093
IUPAC Name 4-hydroxybenzenesulfonamide
SMILES C1=CC(=CC=C1O)S(=O)(=O)N
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10462940 SDS View Documents Acros Organics
433300050
5g Glass bottle £16.50
5g View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 11-06-2020
Please to purchase this item. Need a web account? Register with us today!
10007749 SDS View Documents Acros Organics
433300500
50g Glass bottle £64.20
50g View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 12-06-2020
Please to purchase this item. Need a web account? Register with us today!
10698730 SDS View Documents Acros Organics
433302500
250g Glass bottle £186.80
250g View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 12-06-2020
Please to purchase this item. Need a web account? Register with us today!
Specifications

Chemical Identifiers

1576-43-8
1730.19
DIRCLGLKRZLKHG-UHFFFAOYSA-N
74093
C1=CC(=CC=C1O)S(=O)(=O)N
C6H7NO3S
MFCD00059190
p-hydroxybenzenesulfonamide, p-phenolsulfonamide, benzenesulfonamide, 4-hydroxy, 4-sulfamoylphenol, benzenesulfonamide, p-hydroxy, 4-hydroxyphenylsulfonamide, unii-1iit7r7vpj, p-hydroxybenzenesulphonamide, 4-hydroxybenzenesulphonamide, 4-hydroxybenzene-1-sulfonamide
4-hydroxybenzenesulfonamide

Specifications

4-Hydroxybenzenesulfonamide
Glass bottle
165.4°C to 168.4°C
1576-43-8
100.0
C6H7NO3S
MFCD00059190
DIRCLGLKRZLKHG-UHFFFAOYSA-N
4-hydroxybenzenesulfonamide
74093
Crystalline Powder
Authentic
Yellow
5g
96.0
96% min. (HPLC)
HOC6H4SO2NH2
p-hydroxybenzenesulfonamide, p-phenolsulfonamide, benzenesulfonamide, 4-hydroxy, 4-sulfamoylphenol, benzenesulfonamide, p-hydroxy, 4-hydroxyphenylsulfonamide, unii-1iit7r7vpj, p-hydroxybenzenesulphonamide, 4-hydroxybenzenesulphonamide, 4-hydroxybenzene-1-sulfonamide
C1=CC(=CC=C1O)S(=O)(=O)N
1730.19
1730.19
97%
Safety and Handling

Safety and Handling

Product Identifier
  • 4-Hydroxybenzenesulfonamide

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.