missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxy-6-(trifluoromethyl)pyrimidine-2-thiol, 97+%, Thermo Scientific™

Catalog Number p-7059056
Click to view available options
Quantity:
0.5 g
2 g
10 g
This item is not returnable. View return policy
368-54-7
C5H3F3N2OS
196.147
MFCD00112978
YZAOETRYQWFEOY-UHFFFAOYSA-N
4-hydroxy-6-trifluoromethyl pyrimidine-2-thiol, 2-mercapto-6-trifluoromethyl pyrimidin-4-ol, 2-sulfanyl-6-trifluoromethyl pyrimidin-4-ol, 2-sulfanyl-6-trifluoromethyl-4-pyrimidinol, 4-hydroxy-2-mercapto-6-trifluoromethyl pyrimidine, 6-trifluoromethyl-2-thiouracil, 2-thioxo-6-trifluoromethyl-2,3-dihydropyrimidin-4 1h-one, 4-hydroxy-2-mercapto-6-trifluoromethylpyrimidine, uracil, 2-thio-6-trifluoromethyl, 2-mercapto-6-trifluoromethyl-pyrimidin-4-ol
1239796
2-sulfanylidene-6-(trifluoromethyl)-1H-pyrimidin-4-one
C1=C(NC(=S)NC1=O)C(F)(F)F
This item is not returnable. View return policy
368-54-7
C5H3F3N2OS
196.147
MFCD00112978
YZAOETRYQWFEOY-UHFFFAOYSA-N
4-hydroxy-6-trifluoromethyl pyrimidine-2-thiol, 2-mercapto-6-trifluoromethyl pyrimidin-4-ol, 2-sulfanyl-6-trifluoromethyl pyrimidin-4-ol, 2-sulfanyl-6-trifluoromethyl-4-pyrimidinol, 4-hydroxy-2-mercapto-6-trifluoromethyl pyrimidine, 6-trifluoromethyl-2-thiouracil, 2-thioxo-6-trifluoromethyl-2,3-dihydropyrimidin-4 1h-one, 4-hydroxy-2-mercapto-6-trifluoromethylpyrimidine, uracil, 2-thio-6-trifluoromethyl, 2-mercapto-6-trifluoromethyl-pyrimidin-4-ol
1239796
2-sulfanylidene-6-(trifluoromethyl)-1H-pyrimidin-4-one
C1=C(NC(=S)NC1=O)C(F)(F)F

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 251°C
Quantity 0.5 g
Formula Weight 196.1
Percent Purity ≥97%
Physical Form Powder
Chemical Name or Material 4-Hydroxy-6- (trifluoromethyl)pyrimidine-2-thiol
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'