missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxy-6-methyl-3-nitro-2-pyridone, 97%, Thermo Scientific Chemicals

Catalog Number p-7024649
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
4966-90-9
C6H6N2O4
170.12
MFCD00167612
QIKWTNPFTOEELW-UHFFFAOYSA-N
4-hydroxy-6-methyl-3-nitro-2-pyridone, 4-hydroxy-6-methyl-3-nitropyridin-2 1h-one, 6-methyl-3-nitropyridine-2,4-diol, 2,4-dihydroxy-6-methyl-3-nitropyridine, 2-hydroxy-6-methyl-3-nitro-1h-pyridin-4-one, 4-hydroxy-6-methyl-3-nitro-1,2-dihydropyridin-2-one, 4-hydroxy-6-methyl-3-nitro-2 1h-pyridinone, 4-hydroxy-6-methyl-3-nitrohydropyridin-2-one, pubchem3892, acmc-209khe
54685619
4-hydroxy-6-methyl-3-nitro-1H-pyridin-2-one
CC1=CC(=O)C(=C(O)N1)[N+]([O-])=O
This item is not returnable. View return policy
4966-90-9
C6H6N2O4
170.12
MFCD00167612
QIKWTNPFTOEELW-UHFFFAOYSA-N
4-hydroxy-6-methyl-3-nitro-2-pyridone, 4-hydroxy-6-methyl-3-nitropyridin-2 1h-one, 6-methyl-3-nitropyridine-2,4-diol, 2,4-dihydroxy-6-methyl-3-nitropyridine, 2-hydroxy-6-methyl-3-nitro-1h-pyridin-4-one, 4-hydroxy-6-methyl-3-nitro-1,2-dihydropyridin-2-one, 4-hydroxy-6-methyl-3-nitro-2 1h-pyridinone, 4-hydroxy-6-methyl-3-nitrohydropyridin-2-one, pubchem3892, acmc-209khe
54685619
4-hydroxy-6-methyl-3-nitro-1H-pyridin-2-one
CC1=CC(=O)C(=C(O)N1)[N+]([O-])=O

It is employed as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as a pharmaceutical intermediate.

Solubility
Solubility in N,N-DMF gives very faint turbidity.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY
Melting Point >250°C (decomposition)
Quantity 5 g
Beilstein 1529425
Formula Weight 170.13
Percent Purity 97%
Chemical Name or Material 4-Hydroxy-6-methyl-3-nitro-2-pyridone

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'