missing translation for 'onlineSavingsMsg'
Learn More

4'-Hydroxy-4-biphenylcarbonitrile, 95%, Thermo Scientific™

Catalog Number p-144544
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
19812-93-2
MFCD00059625
ZRMIETZFPZGBEB-UHFFFAOYSA-N
4-cyano-4'-hydroxybiphenyl, 4'-hydroxy-4-biphenylcarbonitrile, 4-4-hydroxyphenyl benzonitrile, 4'-hydroxybiphenyl-4-carbonitrile, 4'-cyano-4-hydroxybiphenyl, 4'-hydroxy-1,1'-biphenyl-4-carbonitrile, 4'-cyano-4-biphenylol, 4-cyano-4'-hydroxy-biphenyl, 4'-hydroxy-4-cyanobiphenyl, 4-hydroxy-4-biphenylcarbonitrile
140610
4-(4-hydroxyphenyl)benzonitrile
C1=CC(=CC=C1C#N)C2=CC=C(C=C2)O
This item is not returnable. View return policy
19812-93-2
MFCD00059625
ZRMIETZFPZGBEB-UHFFFAOYSA-N
4-cyano-4'-hydroxybiphenyl, 4'-hydroxy-4-biphenylcarbonitrile, 4-4-hydroxyphenyl benzonitrile, 4'-hydroxybiphenyl-4-carbonitrile, 4'-cyano-4-hydroxybiphenyl, 4'-hydroxy-1,1'-biphenyl-4-carbonitrile, 4'-cyano-4-biphenylol, 4-cyano-4'-hydroxy-biphenyl, 4'-hydroxy-4-cyanobiphenyl, 4-hydroxy-4-biphenylcarbonitrile
140610
4-(4-hydroxyphenyl)benzonitrile
C1=CC(=CC=C1C#N)C2=CC=C(C=C2)O
CAS Min % 94.0
CAS Max % 100.0
Melting Point 192°C to 199°C
Color Yellow to Beige
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Molecular Formula C13H9NO
Linear Formula NC(C6H4)2OH
Quantity 1 g
Solubility Information Solubility in water: .. Other solubilities: soluble in methanol and acetone
Molecular Weight (g/mol) 195.22
Formula Weight 195.22
Percent Purity 95%
Physical Form Crystalline Powder
Chemical Name or Material 4′-Hydroxy-4-biphenylcarbonitrile, 95%
Show More Show Less
compliance-icons
Product Identifier
  • 4'-Hydroxy-4-biphenylcarbonitrile
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'