missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxy-3-(trifluoromethyl)benzoic acid, 97%, Thermo Scientific™

Catalog Number 13488638
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Glass bottle
This item is not returnable. View return policy
220239-68-9
C8H5F3O3
206.12
MFCD01091008
DPVRVZQEDJVWLS-UHFFFAOYSA-N
4-hydroxy-3-trifluoromethyl benzoic acid, benzoic acid, 4-hydroxy-3-trifluoromethyl, 4-hydroxy-3-trifluoromethyl benzoicacid, acmc-1cdpd, ksc495o6p, 4-hydroxy-3-trifluoromethyl-benzoic acid, 4-oxidanyl-3-trifluoromethyl benzoic acid
2775112
4-hydroxy-3-(trifluoromethyl)benzoic acid
OC(=O)C1=CC(=C(O)C=C1)C(F)(F)F
This item is not returnable. View return policy
220239-68-9
C8H5F3O3
206.12
MFCD01091008
DPVRVZQEDJVWLS-UHFFFAOYSA-N
4-hydroxy-3-trifluoromethyl benzoic acid, benzoic acid, 4-hydroxy-3-trifluoromethyl, 4-hydroxy-3-trifluoromethyl benzoicacid, acmc-1cdpd, ksc495o6p, 4-hydroxy-3-trifluoromethyl-benzoic acid, 4-oxidanyl-3-trifluoromethyl benzoic acid
2775112
4-hydroxy-3-(trifluoromethyl)benzoic acid
OC(=O)C1=CC(=C(O)C=C1)C(F)(F)F
Melting Point 144.0°C to 147.0°C
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Linear Formula CF3C6H3CO2H(OH)
Quantity 250 mg
Formula Weight 206.12
Percent Purity 97%
Chemical Name or Material 4-Hydroxy-3-(trifluoromethyl)benzoic acid
compliance-icons
Product Identifier
  • 4-Hydroxy-3-(trifluoromethyl)benzoic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'