missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxy-3-methoxybenzyl alcohol, 99%, Thermo Scientific Chemicals

Catalog Number p-145497
19.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.


Click to view available options
Quantity:
50 g
250 g
1 kg
Packaging:
Plastic bottle
This item is not returnable. View return policy
498-00-0
C8H10O3
154.17
MFCD00004659
ZENOXNGFMSCLLL-UHFFFAOYSA-N
vanillyl alcohol, 4-hydroxy-3-methoxybenzyl alcohol, 4-hydroxymethyl-2-methoxyphenol, vanillin alcohol, vanillic alcohol, 4-hydroxy-3-methoxybenzenemethanol, benzenemethanol, 4-hydroxy-3-methoxy, vanillol, 4-hydroxy-3-methoxybenzylalcohol, 4-hydroxy-3-methoxyphenylmethanol
62348
CHEBI:18353
4-(hydroxymethyl)-2-methoxyphenol
COC1=CC(CO)=CC=C1O
This item is not returnable. View return policy
498-00-0
C8H10O3
154.17
MFCD00004659
ZENOXNGFMSCLLL-UHFFFAOYSA-N
vanillyl alcohol, 4-hydroxy-3-methoxybenzyl alcohol, 4-hydroxymethyl-2-methoxyphenol, vanillin alcohol, vanillic alcohol, 4-hydroxy-3-methoxybenzenemethanol, benzenemethanol, 4-hydroxy-3-methoxy, vanillol, 4-hydroxy-3-methoxybenzylalcohol, 4-hydroxy-3-methoxyphenylmethanol
62348
CHEBI:18353
4-(hydroxymethyl)-2-methoxyphenol
COC1=CC(CO)=CC=C1O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point 112.0°C to 115.0°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Plastic bottle
Linear Formula HOC6H3(OCH3)CH2OH
Quantity 50 g
Beilstein 06,IV,7381
Formula Weight 154.16
Percent Purity 99%
Physical Form Fine Crystalline Powder
Chemical Name or Material 4-Hydroxy-3-methoxybenzyl alcohol, 99%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'