missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 4-Hydroxy-2-methylbenzeneboronic acid, 98%

Catalog Number p-7035351
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
493035-82-8
C7H9BO3
151.96
MFCD03788424
OYIYNIONWDBJIF-UHFFFAOYSA-N
4-hydroxy-2-methylbenzeneboronic acid, 4-hydroxy-2-methyl phenylboronic acid, 4-borono-3-methylphenol, 4-hydroxy-2-methylphenyl boronic acid, 4-hydroxy-2-methyl phenyl boronic acid, 2-borono-5-hydroxytoluene, boronic acid, 4-hydroxy-2-methylphenyl, 5-hydroxytoluene-2-boronic acid, 4-hydroxy-2-methylphenylboronicacid, tavaborole impurity 3
2773448
(4-hydroxy-2-methylphenyl)boronic acid
CC1=CC(O)=CC=C1B(O)O
This item is not returnable. View return policy
493035-82-8
C7H9BO3
151.96
MFCD03788424
OYIYNIONWDBJIF-UHFFFAOYSA-N
4-hydroxy-2-methylbenzeneboronic acid, 4-hydroxy-2-methyl phenylboronic acid, 4-borono-3-methylphenol, 4-hydroxy-2-methylphenyl boronic acid, 4-hydroxy-2-methyl phenyl boronic acid, 2-borono-5-hydroxytoluene, boronic acid, 4-hydroxy-2-methylphenyl, 5-hydroxytoluene-2-boronic acid, 4-hydroxy-2-methylphenylboronicacid, tavaborole impurity 3
2773448
(4-hydroxy-2-methylphenyl)boronic acid
CC1=CC(O)=CC=C1B(O)O
Melting Point 100°C to 104°C
Quantity 250 mg
Formula Weight 151.96
Percent Purity 98%
Chemical Name or Material 4-Hydroxy-2-methylbenzeneboronic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'