missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxy-2-methoxybenzaldehyde, 98%, ACROS Organics™

Brand:  Acros Organics 335930250

Code : 29

Additional Details : CAS Number : 18278-34-7 Weight : 0.07500kg


Quantity 25g
Packaging Glass bottle

 View more versions of this product

Product Code. 11904801

  • £244.50 / 25g


Estimated Shipment
22-06-2018
Add to basket


Specifications

Chemical Identifiers

4-Hydroxy-2-methoxybenzaldehyde
97.5
98%
C8H8O3
152.15
18278-34-7
100.0
98%
CH(O)C6H3(OH)OCH3

Specifications

4-Hydroxy-2-methoxybenzaldehyde
153.0° to 158.0°C
18278-34-7
100.0
Glass bottle
98%
C8H8O3
152.15
Off-white to light brown
25g
97.5
Authentic
Powder
98%
CH(O)C6H3(OH)OCH3
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 4-Hydroxy-2-methoxybenzaldehyde

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS