missing translation for 'onlineSavingsMsg'
Learn More

4-Hydroxy-1H-pyrazolo[3,4-d]pyrimidine, 98%, Thermo Scientific Chemicals

Catalog Number p-7023426
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
315-30-0
C5H4N4O
136.11
MFCD00599413
OFCNXPDARWKPPY-UHFFFAOYSA-N
allopurinol, 1h-pyrazolo 3,4-d pyrimidin-4-ol, zyloprim, zyloric, lopurin, atisuril, bleminol, caplenal, uripurinol, embarin
2094
CHEBI:40279
1,2-dihydropyrazolo[3,4-d]pyrimidin-4-one
O=C1N=CN=C2NNC=C12
This item is not returnable. View return policy
315-30-0
C5H4N4O
136.11
MFCD00599413
OFCNXPDARWKPPY-UHFFFAOYSA-N
allopurinol, 1h-pyrazolo 3,4-d pyrimidin-4-ol, zyloprim, zyloric, lopurin, atisuril, bleminol, caplenal, uripurinol, embarin
2094
CHEBI:40279
1,2-dihydropyrazolo[3,4-d]pyrimidin-4-one
O=C1N=CN=C2NNC=C12

Xanthine oxidase inhibitor; decreases uric acid production

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point >300°C
Odor Odorless
Quantity 5 g
UN Number UN2811
Formula Weight 136.11
Percent Purity 98%
Chemical Name or Material 4-Hydroxy-1H-pyrazolo[3,4-d]pyrimidine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'