missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Hydroxy-1-indanone, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
4-Hydroxy-1-indanone is a hydroxylated indanone used for synthesis. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuffs.
Solubility
Soluble in ethanol. (25mg/ml)
Notes
Incompatible with oxidizing agents. Store at room temperature. Avoid excessive heat and light.
4-Hydroxy-1-indanone is a hydroxylated indanone used for synthesis. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuffs.
Solubility
Soluble in ethanol. (25mg/ml)
Notes
Incompatible with oxidizing agents. Store at room temperature. Avoid excessive heat and light.
Specifications
Melting Point | 242°C to 244°C |
Quantity | 1 g |
Solubility Information | Soluble in ethanol. (25mg/ml) |
Formula Weight | 148.16 |
Percent Purity | 97% |
Chemical Name or Material | 4-Hydroxy-1-indanone |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'