missing translation for 'onlineSavingsMsg'
Learn More

4-Hydrazinobenzoic acid hydrochloride, 97%, Acros Organics

Brand:  Acros Organics 450670500

Code : NEW

Additional Details : CAS Number : 24589-77-3 Weight : 0.05000kg


Quantity 50g

 View more versions of this product

Product Code. 15139323

  • £92.50 / 50g


EU Stock 2
Add to basket


Specifications

Chemical Identifiers

4-Hydrazinobenzoic acid hydrochloride
97%
C7H8N2O2·HCl
188.61

Specifications

4-Hydrazinobenzoic acid hydrochloride
50g
97%
C7H8N2O2·HCl
188.61
252.0° to 254.0°C
24589-77-3
97%
Glass bottle
Solubility in water: soluble.
Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 4-Hydrazinobenzoic acid hydrochloride

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.