missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Hydrazinobenzene-1-sulfonamide hydrochloride, Maybridge™
Click to view available options
Quantity:
10 g
50 g
Specifications
CAS Min % | 94.5 |
CAS Max % | 100.0 |
Assay Percent Range | 95% |
Packaging | Amber Glass Bottle |
Linear Formula | NH2NHC6H4SO2NH2·HCl |
Quantity | 10 g |
Formula Weight | 223.68 |
Percent Purity | 95% |
Chemical Name or Material | 4-Hydrazinobenzene-1-sulfonamide hydrochloride |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'