Learn More
Justrite™ 4 Gallon Sure-Grip™ EX Countertop Corrosives/Acid Steel Safety Cabinet With 1 Self-Closing Door
Increase workplace safety and improve cleaniliness with the proper safety and storage equipment. The Justrite™ 4 Gallon Sure-Grip™ EX Countertop Corrosives/Acid Steel Safety Cabinet With 1 Self-Closing Door is specially designed for safely storing acids and corrosives in laboratories.
Brand:  Justrite™ 8904221
Code : 03
Additional Details : Weight : 29.49000kg
Product Code. 11965416
  • £1065.00 / Each
Estimated Shipment: 14-08-2024
to see stock.
Description
Description
Countertop and Compac Cabinets let you customize your work areas and make the most of available space. Whether adding to existing storage space or making chemicals readily available at workstation locations, these smaller-sized cabinets are the solution when space is at a premium.
Specifications
Specifications
17 x 17 x 22 in.
4 gallon safety cabinet with self-closing door; 18 Ga CR Steel
Safety Cabinet
Steel (CR)
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
View accessory products