missing translation for 'onlineSavingsMsg'
Learn More

4-Formylphenylboronic acid pinacol ester, 97%, Thermo Scientific Chemicals

Catalog Number p-4602034
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
128376-64-7
C13H17BO3
232.09
MFCD04972375
DMBMXJJGPXADPO-UHFFFAOYSA-N
4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzaldehyde, 4-formylphenylboronic acid pinacol ester, 4-formylbenzeneboronic acid, pinacol ester, 4-formylphenylboronic acid pinacolate, 4-formylphenylboronic acid pinacol cyclic ester, 4-formylphenylboronic acid, pinacol ester, 4-tetramethyl-1,3,2-dioxaborolan-2-yl benzaldehyde, 4-formylbenzeneboronic acid pinacol ester, benzaldehyde, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 4-formylphenylboronic acid, pinacol cyclic ester
2769536
4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzaldehyde
CC1(C)OB(OC1(C)C)C1=CC=C(C=O)C=C1
This item is not returnable. View return policy
128376-64-7
C13H17BO3
232.09
MFCD04972375
DMBMXJJGPXADPO-UHFFFAOYSA-N
4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzaldehyde, 4-formylphenylboronic acid pinacol ester, 4-formylbenzeneboronic acid, pinacol ester, 4-formylphenylboronic acid pinacolate, 4-formylphenylboronic acid pinacol cyclic ester, 4-formylphenylboronic acid, pinacol ester, 4-tetramethyl-1,3,2-dioxaborolan-2-yl benzaldehyde, 4-formylbenzeneboronic acid pinacol ester, benzaldehyde, 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 4-formylphenylboronic acid, pinacol cyclic ester
2769536
4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzaldehyde
CC1(C)OB(OC1(C)C)C1=CC=C(C=O)C=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 59.0°C to 63.0°C
Assay Percent Range 96% min. (GC)
Quantity 5 g
Formula Weight 232.09
Percent Purity 97%
Chemical Name or Material 4-Formylphenylboronic acid pinacol ester
compliance-icons
Product Identifier
  • 4-Formylphenylboronic acid pinacol ester
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'