missing translation for 'onlineSavingsMsg'
Learn More

4-Fluorophenylacetyl chloride, 98%, ACROS Organics™

£26.42 - £423.00

Chemical Identifiers

CAS 459-04-1
Molecular Formula C8H6ClFO
Molecular Weight (g/mol) 172.59
MDL Number MFCD00674181
InChI Key SIOJFYRPBYGHOO-UHFFFAOYSA-N
Synonym 4-fluorophenylacetyl chloride, 2-4-fluorophenyl acetyl chloride, 4-fluorophenylacetylchloride, 4-fluorobenzeneacetyl chloride, p-fluorophenylacetyl chloride, benzeneacetyl chloride, 4-fluoro, 2-4-fluorophenyl ethanoyl chloride, 4-fluoro phenylacetyl chloride, pubchem17517, acmc-1aivq
PubChem CID 3744988
IUPAC Name 2-(4-fluorophenyl)acetyl chloride
SMILES C1=CC(=CC=C1CC(=O)Cl)F
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10177532
SDS View Documents
Acros Organics
376120010
1g Glass bottle
£26.42
1g
View alternative products
Save Up to 
»
In stock, ready to ship: 1 Please to purchase this item. Need a web account? Register with us today!
10013452
SDS View Documents
Acros Organics
376120050
5g Glass bottle
£85.09
5g
View alternative products
Save Up to 
»
In stock, ready to ship: 1 Please to purchase this item. Need a web account? Register with us today!
10721145
SDS View Documents
Acros Organics
376120250
25g Glass bottle
£423.00
25g
View alternative products
Save Up to 
»
Estimated Shipment
15-12-2020
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

459-04-1
172.59
SIOJFYRPBYGHOO-UHFFFAOYSA-N
3744988
C1=CC(=CC=C1CC(=O)Cl)F
C8H6ClFO
MFCD00674181
4-fluorophenylacetyl chloride, 2-4-fluorophenyl acetyl chloride, 4-fluorophenylacetylchloride, 4-fluorobenzeneacetyl chloride, p-fluorophenylacetyl chloride, benzeneacetyl chloride, 4-fluoro, 2-4-fluorophenyl ethanoyl chloride, 4-fluoro phenylacetyl chloride, pubchem17517, acmc-1aivq
2-(4-fluorophenyl)acetyl chloride

Specifications

4-Fluorophenylacetyl chloride
107°C
Glass bottle
1.259
Yellow
459-04-1
100.0
C8H6ClFO
MFCD00674181
Solubility in water: reacts
C1=CC(=CC=C1CC(=O)Cl)F
172.59
172.59
98%
1.2590g/mL
Authentic
1.5110 to 1.513
60°C (8.0mmHg)
1g
97.5
97.5% min. (GC)
FC6H4CH2C(O)Cl
4-fluorophenylacetyl chloride, 2-4-fluorophenyl acetyl chloride, 4-fluorophenylacetylchloride, 4-fluorobenzeneacetyl chloride, p-fluorophenylacetyl chloride, benzeneacetyl chloride, 4-fluoro, 2-4-fluorophenyl ethanoyl chloride, 4-fluoro phenylacetyl chloride, pubchem17517, acmc-1aivq
SIOJFYRPBYGHOO-UHFFFAOYSA-N
2-(4-fluorophenyl)acetyl chloride
3744988
Liquid
Safety and Handling

Safety and Handling

Product Identifier
  • 4-Fluorophenylacetyl chloride

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B

Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • EUH014-Reacts violently with water.
  • EUH029-Contact with water liberates toxic gas.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402+P404-Store in a dry place. Store in a closed container.

SDS