missing translation for 'onlineSavingsMsg'
Learn More

4-Fluorophenylacetic acid, 98%, Thermo Scientific Chemicals

Catalog Number p-3758003
Click to view available options
Quantity:
10 g
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
405-50-5
C8H7FO2
154.14
MFCD00004343
MGKPFALCNDRSQD-UHFFFAOYSA-N
4-fluorophenylacetic acid, 2-4-fluorophenyl acetic acid, p-fluorophenylacetic acid, 4-fluorophenyl acetic acid, benzeneacetic acid, 4-fluoro, 4-fluorobenzeneacetic acid, p-fluorophenyl acetic acid, acetic acid, p-fluorophenyl, unii-s5yjv7caz1, 4-fluoro-phenyl-acetic acid
9837
2-(4-fluorophenyl)acetic acid
C1=CC(=CC=C1CC(=O)O)F
This item is not returnable. View return policy
405-50-5
C8H7FO2
154.14
MFCD00004343
MGKPFALCNDRSQD-UHFFFAOYSA-N
4-fluorophenylacetic acid, 2-4-fluorophenyl acetic acid, p-fluorophenylacetic acid, 4-fluorophenyl acetic acid, benzeneacetic acid, 4-fluoro, 4-fluorobenzeneacetic acid, p-fluorophenyl acetic acid, acetic acid, p-fluorophenyl, unii-s5yjv7caz1, 4-fluoro-phenyl-acetic acid
9837
2-(4-fluorophenyl)acetic acid
C1=CC(=CC=C1CC(=O)O)F

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Melting Point 85.0°C to 86.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Linear Formula FC6H4CH2CO2H
Quantity 10 g
Merck Index 15, 4209
Formula Weight 154.14
Percent Purity 98%
Physical Form Shiny Crystalline Powder or Flakes
Chemical Name or Material 4-Fluorophenylacetic acid
Show More Show Less
compliance-icons
Product Identifier
  • 4-Fluorophenylacetic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'