missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Fluorophenylacetic acid, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
100 g
Description
4-Fluorophenylacetic acid is used as an intermediate in the production of fluorinated anesthetics.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Fluorophenylacetic acid is used as an intermediate in the production of fluorinated anesthetics.
Solubility
Insoluble in water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
4-Fluorophenylacetic acid is used as an intermediate in the production of fluorinated anesthetics.
Solubility
Insoluble in water.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Specifications
Melting Point | 81°C to 85°C |
Boiling Point | 164°C (2 mmHg) |
Flash Point | >100°C (212°F) |
Odor | Odorless |
Quantity | 5 g |
Beilstein | 972145 |
Merck Index | 14,4177 |
Solubility Information | Insoluble in water. |
Formula Weight | 154.14 |
Percent Purity | 98% |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'