missing translation for 'onlineSavingsMsg'
Learn More

4-Fluorophenyl isocyanide, 95%, Thermo Scientific Chemicals

Catalog Number p-3778658
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
24075-34-1
C7H4FN
121.11
MFCD04117575
FEUHGXVGNQTPGQ-UHFFFAOYSA-N
4-fluorophenyl isocyanide, 4-fluorophenylisocyanide, 1-isocyano-4-fluorobenzene, benzene,1-fluoro-4-isocyano, benzene, 1-fluoro-4-isocyano-9ci
2734801
1-fluoro-4-isocyanobenzene
FC1=CC=C(C=C1)[N+]#[C-]
This item is not returnable. View return policy
24075-34-1
C7H4FN
121.11
MFCD04117575
FEUHGXVGNQTPGQ-UHFFFAOYSA-N
4-fluorophenyl isocyanide, 4-fluorophenylisocyanide, 1-isocyano-4-fluorobenzene, benzene,1-fluoro-4-isocyano, benzene, 1-fluoro-4-isocyano-9ci
2734801
1-fluoro-4-isocyanobenzene
FC1=CC=C(C=C1)[N+]#[C-]

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 94.0
CAS Max % 100.0
Density 1.1900g/mL
Boiling Point 62°C to 68°C (20.0 hPa)
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Linear Formula FC6H4NC
Quantity 1 g
Specific Gravity 1.19
Solubility Information Solubility in water: insoluble
Formula Weight 121.11
Percent Purity 95%
Physical Form Liquid or Solid
Chemical Name or Material 4-Fluorophenyl isocyanide
Show More Show Less
compliance-icons
Product Identifier
  • 4-Fluorophenyl isocyanide
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'