missing translation for 'onlineSavingsMsg'
Learn More

4-Fluorophenyl chloroformate, 98%, Thermo Scientific Chemicals

Catalog Number p-135472
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
38377-38-7
C7H4ClFO2
174.56
MFCD00013257
MSBGPEACXKBQSX-UHFFFAOYSA-N
4-fluorophenyl chloroformate, 4-fluorophenyl carbonochloridate, acmc-1ad3b, 4-fluorophenyl chlorocarbonate, 4-fluoro phenyl chloroformate, chloroformic acid p-fluorophenyl ester, carbonochloridic acid 4-fluorophenyl ester
2733256
(4-fluorophenyl) carbonochloridate
C1=CC(=CC=C1OC(=O)Cl)F
This item is not returnable. View return policy
38377-38-7
C7H4ClFO2
174.56
MFCD00013257
MSBGPEACXKBQSX-UHFFFAOYSA-N
4-fluorophenyl chloroformate, 4-fluorophenyl carbonochloridate, acmc-1ad3b, 4-fluorophenyl chlorocarbonate, 4-fluoro phenyl chloroformate, chloroformic acid p-fluorophenyl ester, carbonochloridic acid 4-fluorophenyl ester
2733256
(4-fluorophenyl) carbonochloridate
C1=CC(=CC=C1OC(=O)Cl)F

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Coupling reagent
TRUSTED_SUSTAINABILITY
CAS Min % 97.5
CAS Max % 100.0
Color Yellow
Density 1.3290g/mL
Flash Point 76°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula ClCO2C6H4F
Refractive Index 1.4923 to 1.4943
Quantity 1 g
Specific Gravity 1.329
Formula Weight 174.56
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 4-Fluorophenyl chloroformate, 98%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'