missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Fluorophenol, 99%, Thermo Scientific Chemicals
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
100 g
500 g
Description
4-Fluorophenol is a fluorinated phenolic compound with various applications as an starting reagent for the synthesis of pharmaceutical goods.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
4-Fluorophenol is a fluorinated phenolic compound with various applications as an starting reagent for the synthesis of pharmaceutical goods.
Solubility
Soluble in water (80 mg/ml at 20°C), methylene dichloride, chloroform and methanol.
Notes
Keep container tightly closed in a dry and well-ventilated place. Store away from strong bases and oxidizing agent.
4-Fluorophenol is a fluorinated phenolic compound with various applications as an starting reagent for the synthesis of pharmaceutical goods.
Solubility
Soluble in water (80 mg/ml at 20°C), methylene dichloride, chloroform and methanol.
Notes
Keep container tightly closed in a dry and well-ventilated place. Store away from strong bases and oxidizing agent.
Specifications
Melting Point | 44°C to 50°C |
Density | 1.22 |
Boiling Point | 185°C to 188°C |
Flash Point | 68°C (154°F) |
Odor | Phenol-like |
Quantity | 25 g |
UN Number | UN2923 |
Beilstein | 1362752 |
Solubility Information | Soluble in water (80mg/ml at 20°C),methylene dichloride,chloroform and methanol. |
Formula Weight | 112.1 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'