missing translation for 'onlineSavingsMsg'
Learn More

4-Fluorophenethylealcohol, 97%, ACROS Organics™

Catalog Number p-3778635
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
7589-27-7
C8H9FO
140.16
MFCD00002898
MWUVGXCUHWKQJE-UHFFFAOYSA-N
2-4-fluorophenyl ethanol, 4-fluorophenethyl alcohol, 4-fluorophenylethanol, benzeneethanol, 4-fluoro, 2-4-fluorophenyl ethan-1-ol, 4-fluorophenethylic alcohol, phenethyl alcohol, p-fluoro, 4-fluorobenzeneethanol, 4fa
82068
2-(4-fluorophenyl)ethan-1-ol
OCCC1=CC=C(F)C=C1
This item is not returnable. View return policy
7589-27-7
C8H9FO
140.16
MFCD00002898
MWUVGXCUHWKQJE-UHFFFAOYSA-N
2-4-fluorophenyl ethanol, 4-fluorophenethyl alcohol, 4-fluorophenylethanol, benzeneethanol, 4-fluoro, 2-4-fluorophenyl ethan-1-ol, 4-fluorophenethylic alcohol, phenethyl alcohol, p-fluoro, 4-fluorobenzeneethanol, 4fa
82068
2-(4-fluorophenyl)ethan-1-ol
OCCC1=CC=C(F)C=C1
CAS Min % 96.0
CAS Max % 100.0
Color Colorless
Density 1.1200g/mL
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula FC6H4CH2CH2OH
Refractive Index 1.5060 to 1.5080 (20°C, 589nm)
Quantity 25 g
Formula Weight 140.16
Percent Purity ≥96.0%
Physical Form Liquid
Chemical Name or Material 4-Fluorophenethyl alcohol
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'