missing translation for 'onlineSavingsMsg'
Learn More

4-Fluorocinnamic acid, 99%, Thermo Scientific Chemicals

Catalog Number 10656442
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
459-32-5
C9H7FO2
166.15
MFCD00004395
ISMMYAZSUSYVQG-ZZXKWVIFSA-N
4-fluorocinnamic acid, e-3-4-fluorophenyl acrylic acid, 2e-3-4-fluorophenyl prop-2-enoic acid, p-fluorocinnamic acid, 3-4-fluorophenyl acrylic acid, e-p-fluorocinnamic acid, trans-4-fluorocinnamic acid, p-fluoro cinnamic acid, 3-4-fluorophenyl prop-2-enoic acid, 2-propenoic acid, 3-4-fluorophenyl-, 2e
1530234
(E)-3-(4-fluorophenyl)prop-2-enoic acid
C1=CC(=CC=C1C=CC(=O)O)F
This item is not returnable. View return policy
459-32-5
C9H7FO2
166.15
MFCD00004395
ISMMYAZSUSYVQG-ZZXKWVIFSA-N
4-fluorocinnamic acid, e-3-4-fluorophenyl acrylic acid, 2e-3-4-fluorophenyl prop-2-enoic acid, p-fluorocinnamic acid, 3-4-fluorophenyl acrylic acid, e-p-fluorocinnamic acid, trans-4-fluorocinnamic acid, p-fluoro cinnamic acid, 3-4-fluorophenyl prop-2-enoic acid, 2-propenoic acid, 3-4-fluorophenyl-, 2e
1530234
(E)-3-(4-fluorophenyl)prop-2-enoic acid
C1=CC(=CC=C1C=CC(=O)O)F

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
CAS Min % 98.5
CAS Max % 100.0
Melting Point 207.0°C to 210.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula FC6H4CH=CHCO2H
Quantity 25 g
Beilstein 09, II, 395
Formula Weight 166.15
Percent Purity 99%
Physical Form Crystalline Solid
Chemical Name or Material 4-Fluorocinnamic acid, 99%
Show More Show Less
compliance-icons
Product Identifier
  • 4-Fluorocinnamic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'