4-Fluorobenzyl Chloride 98%, ACROS Organics™


Overview
Brand: Acros Organics

Manufacturer Part Number: 119431000

100ML 4-Fluorobenzyl chloride, 98%

UNSPSC: 12350000

Code: 29

Additional Details:
Additional Details: CAS Number: 352-11-4 Weight: 0.17070kg Transport: UN number : 3265 Chem class : 1a Pack group : II


Packaging Glass bottle
Quantity 100mL

Product Code. 10570921

Quantity Price
1 £ 44.2 / 100mL
Estimated Shipment date
from Supplier 06-03-2017
Add to basket


Description and Specification

Chemical Identifiers

Chemical Name or Material 4-Fluorobenzyl chloride
Name Note 98%
CAS 352-11-4
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5% (GC)
Assay Percent Range 98%
Molecular Formula C7H6ClF
Linear Formula FC6H4CH2Cl
Formula Weight 144.58
MDL Number MFCD00000913
Synonym α-Chloro-p-fluorotoluene

Specification

Physical Form Powder or Crystals
Assay ≥97.5% (GC)
Formula Weight 144.58
Quantity 100mL
Color Yellow
CAS Min % 97.5
EINECS Number 206-516-4
Linear Formula FC6H4CH2Cl
Health Hazard 3 GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. IF SWALLOWED: Rinse mouth. Do NOT induce vomiting. Immediately call a POISON CENTER or doctor/physician.
CAS 352-11-4
Infrared Spectrum Authentic
Assay Percent Range 98%
MDL Number MFCD00000913
Refractive Index 1.5120 to 1.5140 (20°C, 589nm)
Health Hazard 2 GHS P Statement Causes severe skin burns and eye damage.
Packaging Glass bottle
Name Note 98%
Molecular Formula C7H6ClF
Health Hazard 1 GHS Signal Word: Danger
CAS Max % 100.0
Density 1.2070g/mL
Chemical Name or Material 4-Fluorobenzyl chloride
TSCA TSCA
Synonym α-Chloro-p-fluorotoluene
Documentation

Technical documents

Safety and Handling

GHS Signal Word: Danger

GHS P Statement Causes severe skin burns and eye damage.

GHS P Statement Wear protective gloves/protective clothing/eye protection/face protection. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. IF SWALLOWED: Rinse mouth. Do NOT induce vomiting. Immediately call a POISON CENTER or doctor/physician.