missing translation for 'onlineSavingsMsg'
Learn More

4-Fluorobenzyl bromide, 97%, ACROS Organics™

£13.90 - £43.30

Chemical Identifiers

CAS 459-46-1
Molecular Formula C7H6BrF
Molecular Weight (g/mol) 189.03
MDL Number MFCD00000359
InChI Key NVNPLEPBDPJYRZ-UHFFFAOYSA-N
Synonym 4-fluorobenzyl bromide, 1-bromomethyl-4-fluorobenzene, p-fluorobenzyl bromide, 4-fluorobenzylbromide, benzene, 1-bromomethyl-4-fluoro, alpha-bromo-p-fluorotoluene, alpha-bromo-4-fluorotoluene, 1-bromomethyl-4-fluoro-benzene, 4-bromomethyl-1-fluorobenzene, para-fluorobenzyl bromide
PubChem CID 68021
IUPAC Name 1-(bromomethyl)-4-fluorobenzene
SMILES C1=CC(=CC=C1CBr)F
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10712051 SDS View Documents Acros Organics
197970050
5g Glass bottle £13.90
5g View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 23-12-2019
Please to purchase this item. Need a web account? Register with us today!
10398570 SDS View Documents Acros Organics
197970250
25g Glass bottle £43.30
25g View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 23-12-2019
Please to purchase this item. Need a web account? Register with us today!
Specifications

Chemical Identifiers

459-46-1
189.03
NVNPLEPBDPJYRZ-UHFFFAOYSA-N
68021
C1=CC(=CC=C1CBr)F
C7H6BrF
MFCD00000359
4-fluorobenzyl bromide, 1-bromomethyl-4-fluorobenzene, p-fluorobenzyl bromide, 4-fluorobenzylbromide, benzene, 1-bromomethyl-4-fluoro, alpha-bromo-p-fluorotoluene, alpha-bromo-4-fluorotoluene, 1-bromomethyl-4-fluoro-benzene, 4-bromomethyl-1-fluorobenzene, para-fluorobenzyl bromide
1-(bromomethyl)-4-fluorobenzene

Specifications

4-Fluorobenzyl bromide
>110°C
Glass bottle
1.517
Colorless to Yellow
97%
96.0
97%
FC6H4CH2Br
05, II, 238
NVNPLEPBDPJYRZ-UHFFFAOYSA-N
1-(bromomethyl)-4-fluorobenzene
68021
Liquid
1.5170g/mL
Authentic
1.5464 to 1.5484
85.0°C (15.0 mmHg)
5g
459-46-1
100.0
C7H6BrF
MFCD00000359
4-fluorobenzyl bromide, 1-bromomethyl-4-fluorobenzene, p-fluorobenzyl bromide, 4-fluorobenzylbromide, benzene, 1-bromomethyl-4-fluoro, alpha-bromo-p-fluorotoluene, alpha-bromo-4-fluorotoluene, 1-bromomethyl-4-fluoro-benzene, 4-bromomethyl-1-fluorobenzene, para-fluorobenzyl bromide
C1=CC(=CC=C1CBr)F
189.03
189.03
97%
Safety and Handling

Safety and Handling

Product Identifier
  • 4-Fluorobenzyl bromide

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B

Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.