missing translation for 'onlineSavingsMsg'
Learn More

4-Fluorobenzoylacetonitrile, 97%, Thermo Scientific Chemicals

Catalog Number p-7054090
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
4640-67-9
C9H6FNO
163.15
MFCD00662062
LOJBBLDAJBJVBZ-UHFFFAOYSA-N
4-fluorobenzoylacetonitrile, 3-4-fluorophenyl-3-oxopropanenitrile, p-fluorobenzoylacetonitrile, unii-k1ltb8k2dh, 3-4-fluorophenyl-3-oxopropionitrile, k1ltb8k2dh, 3-4-fluoro-phenyl-3-oxo-propionitrile, benzenepropanenitrile, 4-fluoro-beta-oxo, alpha-cyano-4-fluoroacetophenone, acetonitrile, p-fluorobenzoyl
2783172
3-(4-fluorophenyl)-3-oxopropanenitrile
FC1=CC=C(C=C1)C(=O)CC#N
This item is not returnable. View return policy
4640-67-9
C9H6FNO
163.15
MFCD00662062
LOJBBLDAJBJVBZ-UHFFFAOYSA-N
4-fluorobenzoylacetonitrile, 3-4-fluorophenyl-3-oxopropanenitrile, p-fluorobenzoylacetonitrile, unii-k1ltb8k2dh, 3-4-fluorophenyl-3-oxopropionitrile, k1ltb8k2dh, 3-4-fluoro-phenyl-3-oxo-propionitrile, benzenepropanenitrile, 4-fluoro-beta-oxo, alpha-cyano-4-fluoroacetophenone, acetonitrile, p-fluorobenzoyl
2783172
3-(4-fluorophenyl)-3-oxopropanenitrile
FC1=CC=C(C=C1)C(=O)CC#N

Used as pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used as pharmaceutical intermediates.

Solubility
Soluble in dimethylformamide, dimethyl sulfoxide.

Notes
Keep container tightly closed. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point 84°C to 88°C
Color White to Yellow
Quantity 1 g
UN Number UN3439
Solubility Information Soluble in dimethylformamide,dimethyl sulfoxide.
Formula Weight 163.15
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 4-Fluorobenzoylacetonitrile

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'