missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Fluorobenzoylacetonitrile, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
Description
Used as pharmaceutical intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Used as pharmaceutical intermediates.
Solubility
Soluble in dimethylformamide, dimethyl sulfoxide.
Notes
Keep container tightly closed. Store away from oxidizing agents.
Used as pharmaceutical intermediates.
Solubility
Soluble in dimethylformamide, dimethyl sulfoxide.
Notes
Keep container tightly closed. Store away from oxidizing agents.
Specifications
Melting Point | 84°C to 88°C |
Color | White to Yellow |
Quantity | 5 g |
UN Number | UN3439 |
Solubility Information | Soluble in dimethylformamide,dimethyl sulfoxide. |
Formula Weight | 163.15 |
Percent Purity | 97% |
Physical Form | Solid |
Chemical Name or Material | 4-Fluorobenzoylacetonitrile |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'