missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Fluorobenzoic hydrazide, 96%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
Specifications
CAS Min % | 95.0 |
CAS Max % | 100.0 |
Melting Point | 161°C to 164°C |
Color | Brown to Orange |
Infrared Spectrum | Authentic |
Assay Percent Range | 95% min. (HPLC) |
Packaging | Glass Bottle |
Quantity | 1 g |
Formula Weight | 154.14 |
Percent Purity | 96% |
Show More |
Safety and Handling
Product Identifier
- 4-Fluorobenzoic hydrazide
- Warning
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'