missing translation for 'onlineSavingsMsg'
Learn More

4-Fluorobenzoic hydrazide, 96%, ACROS Organics™

Brand:  Acros Organics 438100250

Code : 29

Additional Details : CAS Number : 456-06-4 Weight : 0.07500kg


Quantity 25g

 View more versions of this product

Product Code. 10351190

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Specifications

Chemical Identifiers

4-Fluorobenzoic hydrazide
95.0
≥95.0%
C7H7FN2O
456-06-4
100.0
96%
154.14

Specifications

4-Fluorobenzoic hydrazide
162° to 166°C
456-06-4
95.0
Crystalline Powder
96%
154.14
Orange to light brown
25g
Authentic
100.0
≥95.0%
C7H7FN2O
Videos
SDS